Learn More
Selleck Chemical LLC BIX 02188 S1530-5mg
SDP
Supplier:  Selleck Chemical LLC S15305MG
Encompass_Preferred
BIX02188 is a selective inhibitor of MEK5 with IC50 of 4 3 nM also inhibits ERK5 catalytic activity with IC50 of 810 nM and does not inhibit closely related kinases MEK1 MEK2 ERK2 and JNK2
Catalog No. 50-310-8140