Learn More
Cayman Chemical BIX01294hydrochLRIde hydra 5mg
SDP
Supplier:  Cayman Chemical 131245 MG
Encompass_Preferred
A selective inhibitor of G9a histone methyltransferase (IC50 = 1.7 μM); less effectively inhibits G9a-like protein (GLP; IC50 = 38 μM) and has no effect on other known histone methyltransferases
Catalog No. 50-295-4190