Learn More
Medchemexpress LLC BIX02188 | 334949-59-6 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1205550MG
Encompass_Preferred
BIX02188 is a potent, MEK5-selective small-molecule inhibitor supplied for research use. It inhibits MEK5 with an IC50 of 4.3 nM and reduces ERK5 catalytic activity (IC50 810 nM). Supplied as a high-purity solid and available as DMSO solutions for biochemical and cell-based studies.
  • Potent MEK5-selective inhibitor (IC50 4.3 nM).
  • Reduces ERK5 catalytic activity at higher concentrations (IC50 810 nM).
  • High reported purity suitable for research applications.
  • Available in multiple solid sizes and DMSO solution formats for assay flexibility.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-34480