Learn More
ATCC BJ hTERT (human foreskin fibroblast, hTERT-immortalized) - frozen vial, ATCC CRL-3627
Supplier:  ATCC CRL3627
Encompass
BJ hTERT is an hTERT-immortalized derivative of the normal human foreskin fibroblast cell line BJ. These cells provide a stable, immortalized human fibroblast model useful for studies of cell cycle regulation, oncogene function, and related cancer research.
  • hTERT-immortalized human foreskin fibroblast cell line suitable for cellular and oncogene research.
  • Adherent growth properties.
  • Biosafety level 2 organism.
  • Recommended culture: EMEM with 10% fetal bovine serum.
  • Cryopreserve in complete culture medium with 5% DMSO; store in liquid nitrogen vapor phase.
  • Supplied frozen; approximately 1.0 x 10^6 cells per vial.
  • Derived from neonate male tissue; expresses hTERT; originated in 1998.
Catalog No. NC3915700