Learn More
Medchemexpress LLC BJE6-106 | 1564249-38-2 | MFCD32197190 | 99.9% | 381.47 g·mol⁻1 | C26H23NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1178005MG
Encompass_Preferred
BJE6-106 is a small-molecule research inhibitor selective for protein kinase C delta (PKCδ), supplied as a 5 mg solid for laboratory use. It exhibits potent PKCδ inhibition and has been reported to induce caspase-dependent apoptosis in preclinical studies.
  • High purity (≈99.9%) suitable for biochemical assays and screening.
  • Selective PKCδ inhibition with reported IC50 of 0.05 μM.
  • Supplied as a 5 mg solid; soluble in DMSO (50 mg/mL) with ultrasonic assistance.
  • Recommended storage for powder: -20°C for long-term stability.
  • Intended for research use only; not for human or diagnostic applications.
Catalog No. 50-002-27751