Learn More
Chemglass Life Sciences Beaker, Griffin, 1000mL, Polypropylene, Graduated
SDP
Supplier:  Chemglass Life Sciences CG110816
Encompass_Preferred
Beaker, Griffin, 1000mL, Polypropylene, Graduated. Low form polypropylene beakers are chemically resistant with printed graduations that show approximated volume. Steam autoclavable at 121°C. Sold in the package quantities as listed.
Catalog No. 50-872-717