Learn More
Selleck Chemical LLC Buparlisib (BKM120) 50mg 944396-07-0 NVP-BKM120
SDP
Supplier:  Selleck Chemical LLC S224750MG
Encompass_Preferred
Buparlisib (BKM120, NVP-BKM120) is a selective PI3K inhibitor of p110?/?/?/? with IC50 of 52 nM/166 nM/116 nM/262 nM in cell-free assays, respectively. Reduced potency against VPS34, mTOR, DNAPK, with little activity to PI4K?. Buparlisib induces apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-870-6