Learn More
Selleck Chemical LLC BL-918 5mg
SDP
Supplier:  Selleck Chemical LLC S08195MG
Encompass_Preferred
BL-918 is a potent, orally active activator of UNC-51-like kinase 1 (ULK1) with EC50 of 24.14 nM and Kd of 0.719 ?M. BL-918 induces cytoprotective autophagy for Parkinson's disease treatment. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3048