Learn More
Agilent Technologies BL21-CODONPLUS-RIL COMP CELLS
Supplier:  Agilent Technologies 230240
Encompass
BL21-CodonPlus -RIL and -RP Competent Cells correct codon bias, dramatically improves expression when codon bias is a problem. Contains RIL strain for AT-rich genomes and proteins from all organisms.
Catalog No. 50-125-350