Learn More
Agilent Technologies BL21 COMPETENT CELLS
Supplier:  Agilent Technologies 200133
Encompass
BL21 Competent Cells 5 x 0.2mL For extremely toxic proteins use with lambda CE6 under the control of T7 RNA polymerase or use with non-T7 polymerase-based systems
Catalog No. 50-125-013