Learn More
INTACT GENOMICS INC BL21(DE3) Chemically Competent Cells 6x50µl
Supplier:  INTACT GENOMICS INC 105106
Encompass
Intact Genomics BL21(DE3) Chemically Competent E. coli Cells are suitable for transformation and routine protein expression.BL21(DE3) Chemically Competent Cells have the following features: Widely used host background T7 Expression Strain Deficient in both lon (1) and ompT proteases Resistant to phage T1 (fhuA2) B StrainProduct Includes and Storage:BL21(DE3) Competent Cells: -80 ºCpUC19 Control DNA: -20 ºCRecovery Medium: 4 ºC
Catalog No. 50-000-82967