Learn More
INTACT GENOMICS INC BL21 Chemically Competent Cells 6x50µl
Supplier:  INTACT GENOMICS INC 104106
Encompass
Intact Genomics BL21 chemically competent cells are suitable for transformation and routine protein expression from non-T7 vectors.BL21 chemically competent cells have the following features: Widely used host background. Routine non-T7 vector expression. Deficient in both lon (1) and ompT proteases. Resistant to phage T1 (fhuA2).Product Includes and Storage: BL21 Competent Cells: -80 ºCpUC19 Control DNA: -20 ºCRecovery Medium: 4 ºC
Catalog No. 50-000-82968