Learn More
Alkali Scientific Western Blot Boxes, 4 9/16 X 4 9/16 X 1 1/4", Min Volume 7mL, Black, 5/pk
SDP
Supplier:  Alkali Scientific BC120015BK
Encompass
Western blot boxes and containers are opaque black for light sensitive samples. They fit nearly any sized blot and feature a hinged lid that easy to open and close. The corners are 90˚ and they have a flatter and more uniform surface. Use less reagents and antibodies with these cost-efficient western blot boxes. When you match the size of the box with your blot you're able to fit more blots on your rocker and shaker allowing for a smaller blot footprint.
Catalog No. 50-233-5829