Learn More
MSC Vermont Gage Black Guard Pin Gage, 0.0765 in., Tool Steel, Minus Class ZZ, 1 ea.
Supplier:  MSC 62263140
Encompass
This Class ZZ pin gage is designed for precise hole size measurement. Constructed from 52100 tool steel, it features a Black Guard treatment that provides an effective barrier against rust and corrosion for an extended operational life.
  • Manufactured from 52100 tool steel with a hardness of 60 to 65 Rc
  • Class ZZ minus tolerance of -0.0002 in. / +0.0000 in.
  • Dimensions include a 2 in. length and a surface finish of 10 microinches
  • Centerless lapped for high precision
  • Features visible wear indication and size information electro-etched on the piece
  • NIST traceable with an included certificate of accuracy
Catalog No. 50-005-14139