Learn More
GA International Blackout Paper Labels 1.5" x 0.75" / 38.1mm x 19.1mm Rectangle White From -54°C to +93°C (-65.2°F to +199.4°F)
Supplier:  GA International BOP10C35WH
Encompass_Preferred
Permanent blackout paper labels with a unique opaque design that allows them to obscure any existing printed information, including serial numbers, barcodes, and text. Perfectly suited for general office use, laboratory, and warehouse applications, these cover-up labels are available in roll format and compatible with all standard thermal-transfer barcode printers.
Catalog No. 50-005-70913