Learn More
GA International Blackout Paper Labels 1.625" x 1.375" / 41.3mm x 34.9mm Rectangle White From -54°C to +93°C (-65.2°F to +199.4°F)
Supplier:  GA International BOP502C32.5WH
Encompass_Preferred
Permanent blackout paper labels with a unique opaque design that allows them to obscure any existing printed information, including serial numbers, barcodes, and text. Perfectly suited for general office use, laboratory, and warehouse applications, these cover-up labels are available in roll format and compatible with all standard thermal-transfer barcode printers.
Catalog No. 50-005-72655