Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATCC Bladder Epithelial Cell Basal Medium, 485 mL (ATCC PCS-420-032)

Supplier:  ATCC PCS420032

Encompass

Bladder Epithelial Cell Basal Medium (ATCC PCS-420-032) is a sterile, phenol red-free, serum-free basal medium formulated for the in vitro culture of human bladder epithelial cells as part of ATCC's Primary Cell Solutions. When prepared as a complete medium by supplementing with the matching growth kit, the formulation supports cell proliferation and plating efficiency without feeder layers or extracellular matrix coatings. For research use only; not for therapeutic, diagnostic, or human/animal consumption.

  • Sterile, phenol red-free formulation.
  • Serum-free basal medium designed for bladder epithelial cells.
  • Requires supplementation with the appropriate bladder epithelial cell growth kit to produce complete medium.
  • Packaged volume: 485 mL.
  • Storage: 2°C to 8°C.
  • Complete media stable for 30 days after preparation with growth kit.

Catalog No. 50-238-2252


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.