Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences MICROFIX™ Blades 22°, 80 mm length, 8 mm height
SDP

Supplier:  Electron Microscopy Sciences 6305804

Encompass_Preferred

These unique blades prove to offer higher quality sections with an elongated cutting life.
Length: 80 mm
Height: 8 mm
Thickness: 0.25 mm
Description: Ultrafine sections. Kidney and lymph nodes.

Catalog No. 50-192-8511


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.