Learn More
Med Vet International Dynarex Medicut Blades, #15, 100/Box
Supplier:  Med Vet International 4135
Encompass_Preferred
Dynarex Medi-Cut Blades are crafted from premium-quality stainless steel for optimal performance. The Blades feature a finely honed cutting edge to allow for precise and effective surgical incisions. The Medi-Cut Blades are sterile and individually foil packaged for optimal hygiene. Dynarex Medi-Cut Blades are high-quality stainless-steel blades designed with a finely honed cutting edge for improved performance and precision. 
Catalog No. 50-283-0541