Learn More
Pendotech Pendotech Ph And Conductivity Flow Cell
Supplier:  Pendotech PFCRUAB
Encompass
Blank for lab scale pH and conductivity flow cell for 12mm OD probes
Catalog No. NC1832009
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.