Learn More
TARGETMOL CHEMICALS INC BLARCAMESINE HYDROCHLORI 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T411110MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Blarcamesine hydrochloride (AVex-73 hydrochloride) is a Sigma-1 Receptor agonist (IC50 860 nM). purity: 99%
Catalog No. 50-257-9960