Learn More
Medchemexpress LLC Blarcamesine hydrochloride | 195615-84-0 | MFCD30146449 | 99.9% | 317.85 g/mol | C19H24ClNO | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10186450MG
Encompass_Preferred
Blarcamesine hydrochloride is the hydrochloride salt of blarcamesine, a small-molecule Sigma-1 receptor agonist supplied as a research-grade solid for in vitro and preclinical studies. It is provided with characterization data and handling guidance for sealed storage; reported activity includes an IC50 ≈ 860 nM at the Sigma-1 receptor.
Catalog No. 50-002-46060