Learn More
Cayman Chemical BlastIcIdIn A 1mg
SDP
Supplier:  Cayman Chemical 255191 MG
Encompass_Preferred
A bacterial metabolite; inhibits aflatoxin production by A. parasiticus without affecting fungal growth (IC50s = 0.25 and 1.6 μM, respectively); decreases ribosomal protein levels and inhibits protein synthesis in S. cerevisiae
Catalog No. 50-301-5658