Learn More
Cayman Chemical BlastIcIdIn S 5mg
SDP
Supplier:  Cayman Chemical 352525 MG
Encompass_Preferred
A bacterial metabolite with antibacterial and fungicidal activity; is active against B. subtilis, S. lutea, E. coli, P. fluorescens, and M. tuberculosis, as well as P. oryzae, S. mali, G. kaki, and G. lacticolor in a broth dilution assay from 5-50 UG/ml
Catalog No. 50-290-9995