Learn More
ECM Bioscience Blasticidin S Deaminase Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience BP1231
Encompass_Preferred
This antibody was affinity purified using the N-terminal Bsd peptide (without carrier). The antibody detects a 13 kDa* protein corresponding to the molecular mass of Bsd on SDS-PAGE immunoblots of 293 cells transfected with a lentiviral vector that expresses the Bsd gene product, but is not detected in mock transfected cells. The immunogen was A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminal region of Blasticidin S Deaminase* from Aspergillus Terreus. Accession #P0C2P0. Applications = WB, E, ICC; Species Reactivity = N/A; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = BSD, blasticidin
Catalog No. 50-172-0716