Learn More
TARGETMOL CHEMICALS INC BLEBBISTATIN 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2155010MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Blebbistatin ((±)-Blebbistatin) is a selective non-muscle myosin II (NMII) inhibitor. It promotes directional migration of corneal endothelial cells (CECs) and accelerates wound healing. Furthermore it preserves cell junctional integrity and barrier function. purity: 99%
Catalog No. 50-257-7214