Learn More
Apexbio Technology LLC (-)-Blebbistatin, 856925-71-8, MFCD08460907, 25 mg
SDP
Supplier:  Apexbio Technology LLC B138725
Encompass_Preferred
MF: C18H16N2O2, MW: 292.33, Chemical Name: (S)-3a-hydroxy-6-methyl-1-phenyl-3,3a-dihydro-1H-pyrrolo(2,3-b)quinolin-4(2H)-one. (-)-Blebbistatin is a cell-permeable non-muscle myosin II ATPases inhibitor with an IC50 range of 2 uM.
Catalog No. 50-101-3166