Learn More
STEMCELL Technologies (-)-Blebbistatin, Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 72402
Encompass
(-)-Blebbistatin is a selective cell-permeable inhibitor of non-muscle myosin II ATPases (Kovács et al.; Straight et al.).  It rapidly and reversibly inhibits Mg-ATPase activity and in vitro motility of non-muscle myosin IIA and IIB for several species (IC₅₀ = 0.5-5.0 μM), while poorly inhibiting smooth muscle myosin (IC₅₀ = 80 μM) (Limouze et al.).MAINTENANCE AND SELF-RENEWAL· Increases human pluripotent stem cell (hPSC) survival and cloning efficiency after dissociation to single cells, downstream of ROCK inhibition (Chen et al.; Ohgushi et al.; Walker et al.; Xu et al.). · Enables hPSC to be cultured on microcarriers without surface coating (Chen et al.)· Inhibits differentiation of human mesenchymal stem cells (McBeath et al.; Engler et al.).
Catalog No. 50-197-5917