Learn More
TARGETMOL CHEMICALS INC BLEOMYCIN SULFATE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T611610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bleomycin Sulfate (Blenoxane) a glycopeptide antibiotic is an anticancer agent for squamous cell carcinomas (SCC). In UT-SCC-19A cells the IC50 of Bleomycin Sulfate is 4 nM. purity: 99%
Catalog No. 50-257-6597