Learn More
Alkali Scientific Bleomycin Sulfate, 15 Mg
SDP
Supplier:  Alkali Scientific BLE002CP
Encompass_Preferred
Bleomycin Sulfate is a DNA cleaving antibiotic used extensively in molecular biology and cell research for inducing controlled DNA damage. It generates free radicals that interact with DNA, leading to strand breaks that are useful for studying repair pathways and cellular responses to genotoxic stress. This property makes it valuable in cancer research, apoptosis studies, and investigations of oxidative damage mechanisms. It is commonly applied in cell culture systems to evaluate drug responses and genomic stability. The reagent provides researchers with a reliable method for inducing DNA damage under controlled laboratory conditions.
Catalog No. 50-006-60673