Learn More
Alkali Scientific Bleomycin Sulfate, 5 Mg
SDP
Supplier:  Alkali Scientific BLE001CP
Encompass_Preferred
Bleomycin Sulfate is an antibiotic compound widely used in research applications for its ability to induce DNA strand breaks. It interacts with DNA to generate free radicals that cause single and double strand cleavage, making it a valuable tool for studying DNA damage and repair mechanisms. Researchers commonly use bleomycin in cell biology, cancer research, and molecular biology experiments to model oxidative stress and genomic instability. It is also used in apoptosis studies and drug response testing. The compound is suitable for use in various in vitro systems where controlled DNA damage is required for experimental analysis.
Catalog No. 50-006-60777