Learn More
Cayman Chemical BleomycInsulfate 10mg
SDP
Supplier:  Cayman Chemical 1387710 MG
Encompass_Preferred
A glycopeptide antitumor antibiotic that produces single and double-strand DNA breaks in tumor cells to interrupt their cell cycle; has been used for the treatment of Hodgkin’s lymphoma in combination with doxorubicin, squamous cell carcinomas, testicular cancer, as well as in animal models of pulmonary fibrosis
Catalog No. 50-295-4253