Learn More
Bellco Glass, Inc. Blind Port Plug Assembly ,10mm For 3,7 & 15L
SDP
Supplier:  Bellco Glass, Inc. 790900058
Encompass_Preferred
Stainless steal plug for sealing off the 10 mm port on the 3L, 7L and 15L metric headplates.. MADE IN THE USA
Catalog No. 50-194-6455