Learn More
Bellco Glass, Inc. Blind Port Plug Assembly , For 6mm Port
SDP
Supplier:  Bellco Glass, Inc. 790900057
Encompass_Preferred
Stainless steal plug for sealing off the 10 mm port on the 3L, 7L and 15L metric headplates.. MADE IN THE USA
Catalog No. 50-194-6454