Learn More
O Berk Company Plastic CT Closure: 38-400 Black Rib Side / Smooth Top Phenolic, 1600/pk
Supplier:  O Berk Company 07010035
Encompass_Preferred
Plastic CT Closure: 38-400 Black Rib Side / Smooth Top Phenolic with LDPE Cone Liner; Size 99.99; Neck Diameter 38.00; Neck Finish 400; Material Phenolic Plastic; Color Black; Liner LDPE Cone; Surface Finish Fib Side/Smooth Top 1600/pk
Catalog No. NC9709595