Learn More
Research Products International Corp Blocking Solution Starter Kit
SDP
Supplier:  Research Products International Corp B500101
Encompass_Preferred
A Combination of three different blocking solutions supplied in convenient small quantities. This starter kit is ideal and economical for selecting a blocking solution best suited for a specific protein application or when western blotting is performed infrequently.
Each blocking solution is supplied in a 100ml bottle.
Contents:
Blotto-Tween 20 Blocking Solution in TBS (with azide)
BSA-Tween 20 Blocking Solution in TBS
Casein Blocking Solution in TBS
Catalog No. 50-223-0428