Learn More
Grainger Block 15 X Hplc Or Cryovial Tubes
Supplier:  Grainger 14N149
Encompass_Preferred
The Grainger 14N149 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and allows use of 1/4-inch drive tools with 3/8-inch drive socket accessories. This adapter measures 1-1/2 inches long and has a black oxide finish to resist corrosion. It is rated up to 20,000 psi working torque.
Catalog No. 50-275-1657