Learn More
Med Vet International Jorvet Equine Hoof Positioning Block, J0492X
Supplier:  Med Vet International JOR492X
Encompass_Preferred
These simple blocks elevate the hoof off the ground for better x-rays of the lower limb including evaluation of any P3 rotation.Lightweight, shatterproof, polycarbonate and wood constructionNon-slip surface covered with special 3M material Radiopaque. Cross hairs provide geometric position indicator on the exposed film.Dual cassette holding slots for lateral and AP viewsHolds all sizes of x-ray cassettesGrid ready has room in cassette slot to hold a grid if deemed necessaryAttractive storage bag 12"L x 8" W x 10" H
Catalog No. 50-004-95848