Learn More
Rockland Immunochemicals Blocking Buffer (2X) for Fluorescent Western Blotting - MB-070-0050
SDP
Supplier:  Rockland Immunochemicals MB0700050
Encompass_Preferred
This product is a 2X concentrated stock solution.  Prepare a 1X working solution by diluting 1 part 2X concentrate with 1 parts TBS or equivalent. This buffer was meticulously prepared using ultra pure reagents dissolved in pharmaceutical grade water (WFI) and consists of a proprietary protein formulation in TRIS buffered saline at pH 7.6 with thimerosal added as an antimicrobial agent.
Catalog No. 50-105-9032