Learn More
Rockland Immunochemicals Blocking Buffer for Fluorescent Western Blotting THIMEROSAL FREE 10-PACK (10 x 500 ml) - MB-070-010TF
SDP
Supplier:  Rockland Immunochemicals MB070010TF
Encompass_Preferred
Optimum performance is achieved using this product undiluted.  However, this blocking solution can often be diluted 1:1 in TBS without a significant loss of performance. This buffer was prepared using ultra pure reagents dissolved in pharmaceutical grade water (WFI) and consists of a proprietary protein formulation in TRIS buffered saline at pH 7.6.
Catalog No. 50-105-9033