Learn More
Rockland Immunochemicals Blocking Buffer for Fluorescent Western Blotting 3-PACK (3 x 500 ml) - MB-070-003
SDP
Supplier:  Rockland Immunochemicals MB070003
Encompass_Preferred
Western Blot Blocking Solution is specifically designed for western blotting using fluorochrome conjugated antibodies. Pure nitrocellulose membrane is recommended for maximum performance. Other membranes, such as PVDF or nitrocellulose embedded in a support can be used, but may generate elevated backgrounds. Protein should be transferred from gel to membrane using standard protocols. Blocking buffer can be used for membrane blocking and to dilute both primary and secondary antibodies. Western Blot blocking buffer is suitable for use with fluorescent western blot imaging systems produced by Bio-Rad Laboratories, GE Healthcare, Alpha Innotech, FujiFilm Life Science, Licor Biosciences, UVP and Syngene.
Catalog No. NC9615429