Learn More
Biotium Blocking Buffers Background Reducers Fluorescence Microscopy Chemical Reagents TrueBlack 1/EA
Supplier:  Biotium 23012B1L
Encompass_Preferred
This buffer is a component of the TrueBlack Background Suppressor System (Permeabilizing) It is an excellent general use non-mammalian blocking buffer with detergent for intracellular immunofluorescence Not recommended for use with HRP (contains azide) 1L bottle
Catalog No. 50-005-01578