Learn More
Alkali Scientific Blocking Solution Starter Kit, Contains 100mL of Each Solution: Blotto Tween 20 Blocking Solution in TBS (with Azide), BSA Tween 20 Blocking Solution in TBS, Casein Blocking Solution in TBS
SDP
Supplier:  Alkali Scientific BLO001CP
Encompass_Preferred
The Blocking Solution Starter Kit includes 100mL of each of three commonly used blocking solutions: Blotto Tween 20 Blocking Solution in TBS (with Azide), BSA Tween 20 Blocking Solution in TBS, and Casein Blocking Solution in TBS. Each solution is pre-formulated and ready to use, providing flexibility in choosing the most appropriate blocker for different immunoassays. The kit is ideal for labs that perform various assays requiring protein blocking to reduce background noise and enhance signal clarity. With multiple formulations, this kit provides a comprehensive solution for protein blocking in applications such as ELISA, Western Blot, and immunohistochemistry.
Catalog No. 50-006-61670