Learn More
Med Vet International Blood Collection Micro Tube, Red Top, 100/Ea Bx
Supplier:  Med Vet International MICROTUBE
Encompass_Preferred
Screw cap closure contains integral O-ring. Skirted conical base and label. Length: 45 mm Active Ingredient clotting activator/ serum
Catalog No. 50-282-9821