Learn More
Magbio Genomics Inc Blood STASIS-DNA/RNA 3mL (200)
SDP
Supplier:  Magbio Genomics Inc BSDR3200
Encompass_Preferred
Blood STASIS - DNA/RNA Blood Collection TubesEvacuated tube used for collection, anti-coagulation, stabilization, transport, and storage of venous whole blood as well as for the preservation of blood genomic DNA and RNA
Catalog No. NC2848075