Learn More
Alkali Scientific Blot Stripping Buffer 4X Solution, 500 Milliliters
SDP
Supplier:  Alkali Scientific OTS002CP
Encompass_Preferred
Blot Stripping Buffer 4X Solution is a powerful reagent for the efficient removal of antibodies or proteins from membranes, making them suitable for reuse in subsequent Western blot experiments. This 500mL bottle provides a large quantity of buffer ideal for labs conducting high-throughput analysis or multiple rounds of probing. The buffer’s 4X concentration ensures that it effectively strips the membrane while maintaining its integrity for future use. It is commonly employed in protein analysis experiments, where stripping and re-probing the same membrane for different target proteins are necessary, offering a cost-effective solution for repeated analysis.
Catalog No. 50-006-61262