Learn More
Bioworld Blotto in TBS, 500 mL
Supplier:  Bioworld 402200281
Encompass_Preferred
Blotto in TBS Blot blocking buffer used to block non-specific binding for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.02% Sodium azide in TBS
Catalog No. 50-255-454