Learn More
Alkali Scientific Blotto Tween 20 Blocking Solution in TBS with Azide, 500 Milliliters
SDP
Supplier:  Alkali Scientific LOT002CP
Encompass_Preferred
Blotto Tween 20 Blocking Solution in TBS with Azide is a pre-prepared blocking reagent commonly used in immunoassays such as Western Blot, ELISA, and immunohistochemistry. The addition of Azide ensures that the solution remains free of microbial contamination during storage. This solution blocks non-specific binding sites, ensuring that antibodies or proteins of interest bind specifically to their targets, enhancing the accuracy of experimental results. Available in a 500-milliliter bottle, it is the perfect size for routine use in laboratories performing protein analysis or other immunological studies, helping to reduce background noise and improve assay sensitivity.
Catalog No. 50-006-60554