Learn More
Cayman Chemical BLT1 Receptrmouse Monclo 100ug
SDP
Supplier:  Cayman Chemical 30455100 UG
Encompass_Preferred
Immunogen: BLT1 receptor-expressing cells • Host: Mouse • Species Reactivity: (+) Mouse; Other species not tested • Cross Reactivity: (+) BLT1 • Applications: FC
Catalog No. 50-299-1664