Learn More
TARGETMOL CHEMICALS INC BLU-222 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T843211MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BLU-222 is an orally available but selective cell cycle protein-dependent kinase (CDK2) inhibitor with antitumor activity for the study of ovarian and uterine cancer. Purity 99.88%
Catalog No. 50-287-9156